missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Oxyclozanide, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 2277-92-1 |
| Physical Form | Neat |
| Linear Formula | C13H6 Cl5 NO3 |
| MDL Number | MFCD00864507 |
| Synonym | 2,3,5-Trichloro-N-(3,5-dichloro-2-hydroxyphenyl)-6-hydroxybenzamide; 3,3 ′,5,5 ′,6-Pentachloro-2 ′-hydroxysalicylanilide |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 401.46 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH