missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Oxolinic acid-(ethyl-d5), MilliporeSigma™ Supelco™
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 1189890-98-9 |
| Physical Form | Neat |
| Linear Formula | C13D5H6 NO5 |
| MDL Number | MFCD22565784 |
| Synonym | 5-(Ethyl-d5)-8-oxo-5,8-dihydro-1,3-dioxolo[4,5-g]quinoline-7-carboxylic acid |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 266.26 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH