missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Oxacillin sodium salt monohydrate, MilliporeSigma™ Supelco™

Catalog No. 111015362
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015362 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015362 Supplier Merck Emd Millipore Supplier No. 46589100MG
Only null left

Chemical structure: ß-lactam

Environmental and Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 7240-38-2
Physical Form Neat
Linear Formula C19H18N3O5 SNa · H2O
MDL Number MFCD00167146
Type Analytical Standard
Quantity 100 mg
Formula Weight 441.43
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH