missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Orbifloxacin, MilliporeSigma™ Supelco™

Catalog No. 111012636
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012636 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012636 Supplier Merck Emd Millipore Supplier No. 34041100MGR

Orbifloxacin is an antibacterial fluoroquinolone derivative,for use in small animals such as dogs and cats.

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 113617-63-3
Physical Form Neat
Linear Formula C19H20F3N3O3
MDL Number MFCD00864858
Synonym 1-Cyclopropyl-5,6,8-trifluoro-1,4-dihydro-7-(cis-3,5-dimethyl-1-piperazinyl)-4-oxo-quinoline-3-carboxylic acid; Orbax
Type Analytical Standard
Quantity 100 mg
Formula Weight 395.38
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH