missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Ofloxacin, MilliporeSigma™ Supelco™
Description
Environmental and Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 82419-36-1 |
| Physical Form | Neat |
| Linear Formula | C18H20 FN3O4 |
| MDL Number | MFCD00226105 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 361.37 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH