missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Nitrofurazone-13C,15N2, MilliporeSigma™ Supelco™

Catalog No. 111012780
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012780 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012780 Supplier Merck Emd Millipore Supplier No. 3251210MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 1217220-85-3
Physical Form Neat
Linear Formula 13 CC5H6O415N2N2
MDL Number MFCD16652529
Synonym 5-Nitro-2-furaldehyde semicarbazone-13 C,15N2
Type Analytical Standard
Quantity 10 mg
Formula Weight 201.12
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH