missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ (±)-Naproxen-(methoxy-d3), MilliporeSigma™ Supelco™

Catalog No. 111015967
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015967 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015967 Supplier Merck Emd Millipore Supplier No. 3210410MG

(+/-)-Naproxen-(methoxy-d3) may be used as a reference standard for the determination of (+/-)-naproxen-(methoxy-d3) in aqueous samples using solid-phase extraction (SPE) followed by ultra performance liquid chromatography-triple quadrupole mass spectrometry (UPLC-QqQ-MS/MS).

Environmental and Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 958293-79-3
Physical Form Neat
Linear Formula C14D3H11O3
MDL Number MFCD22572922
Synonym (RS)-2-[6-(Methoxy-d3)-2-naphthyl]propionic acid; 6-(Methoxy-d3)-alpha-methyl-2-naphthaleneacetic acid
Type Analytical Standard
Quantity 10 mg
Formula Weight 233.28
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH