missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Metronidazole-OH, MilliporeSigma™ Supelco™

Catalog No. 111014272
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014272 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014272 Supplier Merck Emd Millipore Supplier No. 3400710MGR

Metronidazole-OH is a genotoxic, carcinogenic and mutagenic drug, belonging to the class of nitroimidazoles, used as a broad spectrum veterinary drug for the treatment and prevention of certain bacterial and protozoal diseases in farm animals.

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 4812-40-2
Physical Form Neat
Linear Formula C6H9N3O4
MDL Number MFCD01721480
Synonym 1-(2-Hydroxyethyl)-2-hydroxymethyl-5-nitroimidazole; Hydroxymetronidazole; MNZOH
Type Analytical Standard
Quantity 10 mg
Formula Weight 187.15
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH