missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Methimazole, MilliporeSigma™ Supelco™

Catalog No. 111014861
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014861 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014861 Supplier Merck Emd Millipore Supplier No. 46429250MG

Methimazole is a thyreostatic compound, and an antihormone, which is widely used in medicine for the treatment of hyperthyroidism.

Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 60-56-0
Physical Form Neat
Linear Formula C4H6N2 S
MDL Number MFCD00179321
Melting Point 144°C to 147°C
Synonym 1-Methyl-2-imidazolethiol; 2-Mercapto-1-methylimidazole
Type Analytical Standard
Quantity 250 mg
Formula Weight 114.17
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH