missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Mecillinam, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 32887-01-7 |
| Physical Form | Neat |
| Linear Formula | C15H23N3O3 S |
| MDL Number | MFCD00056869 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 325.43 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH