missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Mebendazole-amine, MilliporeSigma™ Supelco™

Catalog No. 111014446
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014446 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014446 Supplier Merck Emd Millipore Supplier No. 3540310MG

missing translation for 'validMainframeMsg'

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 52329-60-9
Physical Form Neat
Linear Formula C14H11N3 O
MDL Number MFCD01529678
Synonym 2-Amino-5-benzoyl-1 H-benzimidazole
Type Analytical Standard
Quantity 10 mg
Formula Weight 237.26
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH