missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Lanoconazole, MilliporeSigma™ Supelco™

Catalog No. 111012292
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012292 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012292 Supplier Merck Emd Millipore Supplier No. 3299125MG

Lanoconazole is a member of the imidazole family and an antifungal agent, which finds applications as an antidermatophytic drug.

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 101530-10-3
Physical Form Neat
Linear Formula C14H10 ClN3S2
MDL Number MFCD00865590
Synonym 2-[4-(2-Chlorophenyl)-1,3-dithiolan-2-ylidene]-2-imidazol-1-yl-acetonitrile
Type Analytical Standard
Quantity 25 mg
Formula Weight 319.83
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH