missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Kasugamycin hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111013429
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013429 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013429 Supplier Merck Emd Millipore Supplier No. 32354100MG
Only null left

Chemical structure: aminoglycoside

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 19408-46-9
Physical Form Neat
Linear Formula C14H25N3O9 · HCl
MDL Number MFCD00058459
Synonym 3-O-[2-Amino-4-[(carboxyiminomethyl)amino]-2,3,4,6-tetradeoxy-D-arabino-hexopyranosyl]-D-chiro-inositol hydrochloride
Type Analytical Standard
Quantity 100 mg
Formula Weight 415.82
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH