missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ HMMNI-d3, MilliporeSigma™ Supelco™
Description
Environmental and Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 1015855-78-3 |
| Physical Form | Neat |
| Linear Formula | C5H4D3N3O3 |
| MDL Number | MFCD09265283 |
| Synonym | 2-Hydroxymethyl-1-methyl-d3-5-nitro-1 H-imidazole |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 160.15 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH