missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Flumequine, MilliporeSigma™ Supelco™

Catalog No. 111014181
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014181 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014181 Supplier Merck Emd Millipore Supplier No. 45735250MG
Only null left

Chemical structure: fluoroquinolone

Agriculture and Environmental

Specifications

Product Line VETRANAL
CAS 42835-25-6
Physical Form Neat
Linear Formula C14H12 FNO3
MDL Number MFCD00079298
Type Analytical Standard
Quantity 250 mg
Formula Weight 261.25
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH