missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Florfenicol amine, MilliporeSigma™ Supelco™

Catalog No. 111015465
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015465 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015465 Supplier Merck Emd Millipore Supplier No. 3249210MG

Florfenicol amine is a polar metabolite of florfenicol.

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 76639-93-5
Physical Form Neat
Linear Formula C10H14 FNO3 S
MDL Number MFCD19704818
Synonym (alphaR)-alpha-[(1 S)-1-Amino-2-fluoroethyl]-4-(methylsulfonyl)benzenemethanol; D-(-)-threo-2-Amino-3-fluoro-1-[4-(methylsulfonyl)phenyl]-1-propanol
Type Analytical Standard
Quantity 10 mg
Formula Weight 247.29
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH