missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Fenbendazole sulfone, MilliporeSigma™ Supelco™

Catalog No. 111014541
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014541 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014541 Supplier Merck Emd Millipore Supplier No. 3254410MG

Chemical structure: benzimidazole

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 54029-20-8
Physical Form Neat
Linear Formula C15H13N3O4 S
MDL Number MFCD00797921
Synonym 5-Benzenesulfonyl-1 H-benzoimidazol-2-yl)carbamic acid methyl ester; Methyl 5-(phenylsulfonyl)-2-benzimidazolecarbamate
Type Analytical Standard
Quantity 10 mg
Formula Weight 331.35
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH