missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Enrofloxacin-d5 hydrochloride, Analytical Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Product Line | VETRANAL |
| Physical Form | Neat |
| Linear Formula | C19D5H17FN3O3 · HCl |
| Synonym | 1-Cyclopropyl-7-(4-ethyl-d5-1-piperazinyl)-6-fluoro-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid hydrochloride |
| Quantity | 10 mg |
| Formula Weight | 400.89 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH