missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Doramectin, MilliporeSigma™ Supelco™

Catalog No. 111019834
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019834 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019834 Supplier Merck Emd Millipore Supplier No. 33993100MGR
Only null left
Agriculture

Specifications

Product Line VETRANAL
CAS 117704-25-3
Physical Form Neat
Linear Formula C50H74O14
MDL Number MFCD00894747
Synonym 25-Cyclohexyl-5-O-demethyl-25-de(1-methylpropyl)avermectin
Type Analytical Standard
Quantity 100 mg
Formula Weight 899.11
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH