missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ DNC-d8, MilliporeSigma™ Supelco™
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 1156508-87-0 |
| Physical Form | Neat |
| Linear Formula | C13D8H2N4O5 |
| MDL Number | MFCD06656252 |
| Synonym | 4,4 ′-Dinitrocarbanilide-d8; N,N′-Bis(4-nitrophenyl)urea-d8; BNPH-d8 |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 310.29 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH