missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Diclofenac-(acetophenyl ring-13C6) sodium salt hemi(nonahydrate), MilliporeSigma™ Supelco™

Catalog No. 111018049
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018049 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018049 Supplier Merck Emd Millipore Supplier No. 3536110MG
Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 1261393-73-0
Physical Form Neat
Linear Formula 13C6C8H10 Cl2 NNaO2 · 4.5H2 O
MDL Number MFCD20264909
Synonym [2-(2,6-Dichlorophenylamino)phenyl13C6 ]acetic acid sodium salt hemi(nonahydrate)
Type Analytical Standard
Quantity 10 mg
Formula Weight 405.16
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH