missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Dexamethasone 21-acetate, MilliporeSigma™ Supelco™
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 1177-87-3 |
| Physical Form | Neat |
| Linear Formula | C24H31 FO6 |
| MDL Number | MFCD00027407 |
| Synonym | 9 alpha-Fluoro-16 alpha-methyl-11 beta,17 alpha,21-trihydroxy-1,4-pregnadiene-3,20-dione-21-acetate; 9 alpha-Fluoro-16 alpha-methylprednisolone-21-acetate; Prednisolone F acetate |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 434.50 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH