missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Clorprenaline, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 3811-25-4 |
| Physical Form | Neat |
| Linear Formula | C11H16 ClNO |
| MDL Number | MFCD00864144 |
| Synonym | 1-(2-Chlorophenyl)-2-isopropylaminoethanol |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 213.70 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH