missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Clioquinol, MilliporeSigma™ Supelco™

Catalog No. 111019841
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019841 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019841 Supplier Merck Emd Millipore Supplier No. 33931100MGR
Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 130-26-7
Physical Form Neat
Linear Formula C9H5 ClINO
MDL Number MFCD00006787
Synonym 5-Chloro-7-iodo-8-quinolinol; 5-Chloro-8-hydroxy-7-iodoquinoline; Clioquinol; Iodochlorhydroxyquin
Type Analytical Standard
Quantity 100 mg
Formula Weight 305.50
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH