missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Clenpenterol hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111014066
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014066 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014066 Supplier Merck Emd Millipore Supplier No. 3282510MG

Clenpenterol hydrochloride is a salt of clenpenterol which is a β2-agonist compound.

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 37158-47-7
Physical Form Neat
Linear Formula C13H20 Cl2N2 O · HCl
MDL Number MFCD11975209
Synonym 1-(4-Amino-3,5-dichlorophenyl)-2-(1,1-dimethylpropylamino)ethanol hydrochloride
Type Analytical Standard
Quantity 10 mg
Formula Weight 327.68
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH