missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Clencyclohexerol hydrochloride, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 1435934-75-0 |
| Physical Form | Neat |
| Linear Formula | C14H20 Cl2N2O2 · HCl |
| MDL Number | MFCD20264921 |
| Synonym | 4-[2-(4-Amino-3,5-dichlorophenyl)-2-hydroxyethylamino]cyclohexanol hydrochloride |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 355.69 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH