missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Citicoline Sodium Salt Hydrate, Analytical Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Product Line | VETRANAL |
| Physical Form | Neat |
| Linear Formula | C14H25N4NaO11P2 · xH2O |
| MDL Number | MFCD10567436 |
| Quantity | 100 mg |
| Formula Weight | 510.31 (anhydrous basis) |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH