missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Ceftiofur hydrochloride, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 103980-44-5 |
| Physical Form | Neat |
| Linear Formula | C19H17N5O7S3 · HCl |
| MDL Number | MFCD04038020 |
| Synonym | (6 R,7 R)-7-{[(2 Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino}-3-{[(2-furanylcarbonyl)thio]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0 ]oct-2-ene-2-carboxylic acid hydrochloride; 3-[(2-Furylcarbonyl)thio]methyl]-7-[2-(2-amino-4-thiazolyl)-2-(methoxyiminoacetamido)-3-cephem-4-carboxylic acid hydrochloride |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 560.02 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH