missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Cefoperazone dihydrate, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 113826-44-1 |
| Physical Form | Neat |
| Linear Formula | C25H27N9O8S2 · 2H2 O |
| MDL Number | MFCD17171280 |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 681.70 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH