missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Carprofen-d3, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 1173019-42-5 |
| Physical Form | Neat |
| Linear Formula | C15D3H9 ClNO2 |
| MDL Number | MFCD09840202 |
| Synonym | 6-Chloro-alpha-methyl-d3-9 H-carbazole-2-acetic acid |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 276.73 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH