missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Canamycin A Disulfate Salt Hydrate Analytical Standard, MilliporeSigma™ Supelco™
$69.06 - $108.63
Specifications
| CAS | 64013-70-3 (Anhydrous) |
|---|---|
| Physical Form | Neat |
| Molecular Formula | C18H36N4O11 · 2H2SO4 · xH2O |
| Linear Formula | C18H36N4O11 · 2H2SO4 · xH2O |
| UN Number | NONH for all modes of transport |
Specifications
| 64013-70-3 (Anhydrous) | |
| C18H36N4O11 · 2H2SO4 · xH2O | |
| NONH for all modes of transport | |
| 680.66 (Anhydrous Basis) |
| Neat | |
| C18H36N4O11 · 2H2SO4 · xH2O | |
| Kanamycin A disulfate salt | |
| Analytical Standard |
VETRANAL is a registered trademark of Sigma-Aldrich Chemie GmbH