missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Canamycin A Disulfate Salt Hydrate Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022527
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022527 10 mg
111022528 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111022527 Supplier Merck Emd Millipore Supplier No. 4635610MG

Specifications

CAS 64013-70-3 (Anhydrous)
Quantity 10 mg
Physical Form Neat
Molecular Formula C18H36N4O11 · 2H2SO4 · xH2O
Linear Formula C18H36N4O11 · 2H2SO4 · xH2O
UN Number NONH for all modes of transport
Synonym Kanamycin A disulfate salt
Formula Weight 680.66 (Anhydrous Basis)
Grade Analytical

VETRANAL is a registered trademark of Sigma-Aldrich Chemie GmbH