missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Bithionol, MilliporeSigma™ Supelco™

Catalog No. 111015987
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015987 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015987 Supplier Merck Emd Millipore Supplier No. 31622250MG
Only null left

Bithionol is a dichlorophenol compound that shows antimicrobial property.1

Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 97-18-7
Physical Form Neat
Linear Formula C12H6 Cl4O2 S
MDL Number MFCD00055727
Synonym 2,2 ′-Thio-bis(4,6-dichlorophenol); Bis(2-hydroxy-3,5-dichlorophenyl) sulfide
Type Analytical Standard
Quantity 250 mg
Formula Weight 356.05
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH