missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Betamethasone, MilliporeSigma™ Supelco™

Catalog No. 111014090
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014090 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014090 Supplier Merck Emd Millipore Supplier No. 34166100MG

Specifications

Product Line VETRANAL
CAS 378-44-9
Physical Form Neat
Linear Formula C22H29 FO5
MDL Number MFCD00062969
Melting Point 235°C to 237°C
Synonym 9 alpha-Fluoro-11 beta,17 alpha,21-trihydroxy-16 beta-methylpregna-1,4-diene-3,20-dione; 9 alpha-Fluoro-16 beta-methyl-11 beta,17 alpha,21-trihydroxy-1,4-pregnadiene-3,20-dione; 9 alpha-Fluoro-16 beta-methylprednisolone
Type Analytical Standard
Quantity 100 mg
Formula Weight 392.46
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH