missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Amoxicillin trihydrate, MilliporeSigma™ Supelco™

Catalog No. 111014890
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014890 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014890 Supplier Merck Emd Millipore Supplier No. 31586250MG

Chemical structure: ß-lactam

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 61336-70-7
Physical Form Neat
Linear Formula C16H19N3O5 S · 3H2 O
MDL Number MFCD00072029
Type Analytical Standard
Quantity 250 mg
Formula Weight 419.45
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH