missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ Albendazole sulfone, MilliporeSigma™ Supelco™

Catalog No. 111015433
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015433 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015433 Supplier Merck Emd Millipore Supplier No. 3539410MG

Albendazole sulfone is a nonactive metabolite of albendazole sulfoxide (ABZSO).

Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 75184-71-3
Physical Form Neat
Linear Formula C12H15N3O4 S
MDL Number MFCD00600775
Synonym Methyl [5-(propylsulfonyl)-1 H-benzimidazol-2-yl]carbamate
Type Analytical Standard
Quantity 10 mg
Formula Weight 297.33
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH