missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ 6-Propyl-2-thiouracil, MilliporeSigma™ Supelco™

Catalog No. 111014411
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014411 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014411 Supplier Merck Emd Millipore Supplier No. 46698250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Analytical standard for clinical testing.

Pharmaceutical (small molecule)

Specifications

Chemical Name or Material 6-Propyl-2-thiouracil
CAS 51-52-5
Product Line VETRANAL
Flash Point 300°C
Physical Form Neat
Linear Formula C7H10N2 OS
Technique Gas Chromatography
MDL Number MFCD00006041
Melting Point 218°C to 220°C
Synonym 2,3-Dihydro-6-propyl-2-thioxo-4(1 H)-pyrimidinone; 4-Hydroxy-2-mercapto-6-propylpyrimidine
Type Analytical Standard
Solvent or Matrix Neat
Industry Type Clinical Testing
Molecular Weight (g/mol) 170.23
Quantity 250 mg
Formula Weight 170.23
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH