missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ 6-Methyl-2-Thiouracil Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. p-7214091
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022529 10 mg
111022530 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111022529 Supplier Merck Emd Millipore Supplier No. 4645310MG
Only null left

Specifications

CAS 56-04-2
Melting Point ∼330°C (decomposition) (literature)
Molecular Formula C5H6N2OS
Linear Formula C5H6N2OS
MDL Number MFCD00006040
Quantity 10 mg
UN Number UN 3077 9/PGIII
Synonym 4-Hydroxy-2-mercapto 6-methylpyrimidine; MZU
Formula Weight 142.18
Grade Analytical Standard
Physical Form Neat
Show More Show Less

VETRANAL is a registered trademark of Sigma-Aldrich Chemie GmbH