missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ 6-Chloro-2,4-diamino-1,3,5-triazine-13C3, MilliporeSigma™ Supelco™

Catalog No. 111012778
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012778 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012778 Supplier Merck Emd Millipore Supplier No. 3223210MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Agriculture

Specifications

Product Line VETRANAL
CAS 1216850-33-7
Physical Form Neat
Linear Formula 13C3H4 ClN5
MDL Number MFCD09840347
Melting Point >320°C
Type Analytical Standard
Quantity 10 mg
Formula Weight 148.53
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH