missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ 5-Hydroxymebendazole, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 60254-95-7 |
| Physical Form | Neat |
| Linear Formula | C16H15N3O3 |
| Synonym | Methyl 5-[(RS)-alpha-hydroxybenzyl]-1 H-benzimidazole-2-carbamate |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 297.31 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH