missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ 2-NP-AMOZ-d5, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 1173097-59-0 |
| Physical Form | Neat |
| Linear Formula | C15H13D5N4O5 |
| MDL Number | MFCD06656261 |
| Synonym | 5-(Morpholinomethyl)-3-(2-nitrobenzylidenamino)-2-oxazolidinone-d5 |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 339.36 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH