missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ 2-NP-AHD, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 623145-57-3 |
| Physical Form | Neat |
| Linear Formula | C10H8N4O4 |
| MDL Number | MFCD04973440 |
| Synonym | 1-(2-Nitrobenzylidenamino)-2,4-imidazolidinedione |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 248.19 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH