missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ 1-Aminohydantoin hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111013815
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013815 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013815 Supplier Merck Emd Millipore Supplier No. 33655100MGR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Analytical reference standard for the determination of veterinary drug residues by LC–MS/MS.

Pharmaceutical (small molecule)

Specifications

Chemical Name or Material 1-Aminohydantoin Hydrochloride
CAS 2827-56-7
Product Line VETRANAL
Physical Form Neat
Linear Formula C3H5N3O2 · HCl
Technique Gas Chromatography
MDL Number MFCD02181047
Melting Point 201°C to 205°C
Type Analytical Standard
Industry Type Toxicology
Molecular Weight (g/mol) 151.55
Quantity 100 mg
Formula Weight 151.55
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH