missing translation for 'onlineSavingsMsg'
Learn More

VETRANAL™ 1-Amino-2,4-imidazolidinedione-13C3, MilliporeSigma™ Supelco™

Catalog No. 111015965
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015965 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015965 Supplier Merck Emd Millipore Supplier No. 3400610MGR
Pharmaceutical (small molecule)

Specifications

Product Line VETRANAL
CAS 957509-31-8
Physical Form Neat
Linear Formula 13C3H5N3O2
MDL Number MFCD07366660
Synonym AHD-13C3
Type Analytical Standard
Quantity 10 mg
Formula Weight 118.07
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH