missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ VetMAX™ Xeno™ Internal Positive Control - LIZ™ Assay

Catalog No. A29766
Encompass
Change view
Click to view available options
No. of Reactions:
100
500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A29766 100
A29768 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29766 Supplier Applied Biosystems™ Supplier No. A29766

The VetMAX™ Xeno™ Internal Positive Control (IPC) - LIZ™ Assay is a primer-probe mix that detects the Xeno internal positive control. The resultant Xeno data is used to determine the validity of diagnostic test results.

The Xeno IPC LIZ assay is introduced during the qPCR preparation step and carried through the animal health PCR workflow.

Features of the VetMAX Xeno IPC - LIZ Assay include:

  • Provides confidence that qPCR test results are accurate and actionable
  • Easily integrates into any workflow
  • Greatly reduces the likelihood of false negatives

The VetMAX Xeno IPC - LIZ Assay comes in a 25X concentration and easily integrates into animal health PCR workflows, regardless of the target assay, master mix, or sample preparation reagents already in place. Coupled with VetMAX™ Xeno™ Internal Positive Control RNA or VetMAX™ Xeno™ Internal Positive Control DNA, our proprietary design offers a verification layer to help ensure the qPCR test results are accurate and actionable by greatly reducing the likelihood of false negatives.

Xeno IPC assays are included in most VetMAX kits and have been successfully benchmarked against millions of genomes including those relevant to animal health. A VetMAX™ Xeno™ Internal Positive Control - VIC™ Assay (Cat. No. A29765) is also available.

Specifications

Content And Storage 1 x 110 μL Xeno LIZ Primer Probe Mix. Store at -5°C to -30°C.
Format Kit
For Use With (Application) Detection and Identification of Animal Disease
Includes Internal Positive Control Assay
Label or Dye LIZ
No. of Reactions 100
Product Line VetMAX, Xeno
Product Type Internal Positive Control
Quantity 100 reactions
Shipping Condition Dry Ice
Species Not Species-Specific
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.