Learn More
Applied Biosystems™ VetMAX™ Xeno™ Internal Positive Control DNA
Description
Xeno IPC DNA is introduced at the nucleic acid isolation/preparation step and carried through the animal health qPCR workflow.
Features of VetMAX Xeno IPC DNA include:
- Provides confidence that qPCR test results are accurate and actionable
- Easily integrates into any workflow
- Greatly reduces the likelihood of false negatives
VetMAX Xeno IPC DNA comes in a concentration of 10,000 copies/μL and easily integrates into animal health PCR workflows, regardless of the target assay, master mix, or sample preparation reagents already in place. Coupled with a Xeno internal positive control assay, our proprietary design offers a verification layer to help ensure the PCR test results are accurate and actionable by greatly reducing the likelihood of false negatives.
Xeno IPC DNA is a component of our licensed VetMAX Gold detection kits, recommended in the American Association of Veterinary Diagnosticians guidelines, and has been successfully benchmarked against millions of genomes including those relevant to animal health. Xeno IPC RNA is also available (Cat. No. A29761).
Specifications
Specifications
| Content And Storage |
|
| Format | Kit |
| For Use With (Application) | Detection and Identification of Animal Disease |
| Includes | Internal Positive Control |
| No. of Reactions | 500 |
| Product Line | VetMAX, Xeno |
| Product Type | Internal Positive Control DNA |
| Quantity | 500 reactions |
| Shipping Condition | Dry Ice |
| Species | Not Species-Specific |
For Research Use Only. Not for use in diagnostic procedures.