missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ VetMAX™ Xeno™ Internal Positive Control DNA

Catalog No. A29764
Encompass
Change view
Click to view available options
No. of Reactions:
100
500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A29764 100
A29762 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29764 Supplier Applied Biosystems™ Supplier No. A29764

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

VetMAX™ Xeno™ Internal Positive Control (IPC) DNA serves as an internal positive control for the DNA purification process and helps monitor for the presence of PCR inhibitors in animal health molecular detection workflows. This is of particular importance when working with sample types that have a high level of inhibitors such as feces.

Xeno IPC DNA is introduced at the nucleic acid isolation/preparation step and carried through the animal health qPCR workflow.

Features of VetMAX Xeno IPC DNA include:

  • Provides confidence that qPCR test results are accurate and actionable
  • Easily integrates into any workflow
  • Greatly reduces the likelihood of false negatives

VetMAX Xeno IPC DNA comes in a concentration of 10,000 copies/μL and easily integrates into animal health PCR workflows, regardless of the target assay, master mix, or sample preparation reagents already in place. Coupled with a Xeno internal positive control assay, our proprietary design offers a verification layer to help ensure the PCR test results are accurate and actionable by greatly reducing the likelihood of false negatives.

Xeno IPC DNA is a component of our licensed VetMAX Gold detection kits, recommended in the American Association of Veterinary Diagnosticians guidelines, and has been successfully benchmarked against millions of genomes including those relevant to animal health. Xeno IPC RNA is also available (Cat. No. A29761).

Specifications

Content And Storage
  • 1 x 1250 μL Nucleic Acid Dilution Solution. Store at -5°C to -30°C.
  • 1 x 250 μL Xeno DNA Control (10,000 copies/μL). Store at -5°C to -30°C.
Format Kit
For Use With (Application) Detection and Identification of Animal Disease
Includes Internal Positive Control
No. of Reactions 100
Product Line VetMAX, Xeno
Product Type Internal Positive Control DNA
Quantity 100 reactions
Shipping Condition Dry Ice
Species Not Species-Specific

For Research Use Only. Not for use in diagnostic procedures.