Learn More
STERIS Vesphene™ III se Disinfectant Cleaner
This Phenolic Disinfectant is the latest generation of high pH disinfectant specifically formulated for the routine disinfection of hard, non-porous surfaces.
$425.25
Specifications
| Product Type | Phenolic Disinfectant |
|---|---|
| Sterility | Non-sterile |
| Concentration | 0.8% v/v (1:128) |
Description
The Vesphene™ III se Disinfectant is specifically formulated for the routine disinfection of hard, non-porous surfaces. At a use dilution of rate of 0.8% v/v (1:128), the Vesphene™ III Disinfectants kill a broad spectrum of common bacteria and fungi. The alkaline Vesphene™ III Disinfectant formulation can be used alone or in rotation with the LpH™ III Disinfectants. These products have an extensive documentation package to meet your validation objectives and are supported by the STERIS™ Technical Services team for individualized application and validation assistance.
How Vesphene™ III se Disinfectant Works
The STERIS™ Vesphene™ III se Phenolic Disinfectant is a liquid concentrate specifically formulated for the disinfection of hard surfaces. The use-dilution can be applied manually or sprayed.
Why Vesphene™ III se Disinfectant?
- Efficacy
- Phenolic active ingredients provide broad spectrum antimicrobial efficacy.
- Productivity
- Formulation includes surfactants for surface cleaning while disinfecting.
- Flexibility
- Ideal for use in a contamination control rotation.
- Reliability
- Complete lot traceability.
- Assurance
- Each lot documented by Certificate of Analysis.
Specifications
| Phenolic Disinfectant | |
| Non-sterile | |
| 0.8% v/v (1:128) |