Learn More
Poltex Vertical Lab Benchtop Organizer
Catalog No. 22000287
Encompass
Change view
Click to view available options
Holds:
3-pocket
Includes:
No bracket
Pipette controller bracket
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Includes
22000287 3-pocket Pipette controller bracket
22000286 3-pocket No bracket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22000287 Supplier Poltex Supplier No. 3VTORGBRK
Get a Poltex Pipette Holder with $1,500 Purchase
View all promotions
Poltex Vertical Lab Benchtop Organizer stacks tips or other lab items vertically to preserve benchtop space
  • Fits most brands of tip boxes
  • Super durable PETG construction
  • Optional pipette controller bracket
Specifications
Holds 3-pocket
Description Benchtop organizer
Diameter (English) 1.125 in.
Dimensions (D x W x H) 4.5 x 4.5 x 12 in.
For Use With (Equipment) PETG
Includes Pipette controller bracket