missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Vertical Clean Bench IV Bar Kit

Catalog No. 10304219
Change view
Click to view available options
Width (English):
6 ft.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
10304219 6 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10304219 Supplier Labconco™ Supplier No. 3721004

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with 6 ft Purifier Vertical Clean Benches and Filtered PCR Enclosures. IV Bar supports intravenous solution bottles and bags. Includes IV bar, mounting hardware, four hangers and instructions for installation.

  • For use with 6 ft. Purifier Vertical Clean Benches and Filtered PCR Enclosures.
  • IV Bar supports intravenous solution bottles and bags.
  • Includes IV bar, mounting hardware, four hangers and instructions for installation. 

Specifications

Width (English) 6 ft.
Product Type Vetical Clean Bench Bar Kit